Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp86D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260108.2 | NP_001247037.1 | 1 | 579 | 80 | |
NM_001275539.1 | NP_001262468.1 | 1 | 579 | 80 | ||
NM_079585.5 | NP_524309.1 | 1 | 579 | 80 | ||
GenBank mRNA | BT004914.1 | - | 1 | 567 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |