Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014713.4 | NP_001014713.1 | 2 | 2092 | 118 | |
NM_001144670.2 | NP_001138142.1 | 2 | 2124 | 118 | ||
NM_001258518.3 | NP_001245447.1 | 3 | 2381 | 118 | ||
NM_080294.5 | NP_525033.1 | 3 | 2381 | 118 | ||
GenBank mRNA | AY051822.1 | - | 3 | 2350 | 118 | |
M21152.1 | - | 3 | 2375 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |