Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SpdS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169298.3 | NP_731384.1 | 2 | 1058 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |