Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382066.1 | NP_001368966.1 | 4 | 1377 | 61 | |
GenBank mRNA | BT044472.1 | - | 4 | 1342 | 61 |
Gene Symbol: SdicC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316649.1 | NP_001303578.1 | 4 | 1351 | 61 | |
GenBank mRNA | BT150243.1 | - | 4 | 1351 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |