Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Octbeta3R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038954.2 | NP_001034043.2 | 3 | 1543 | 78 | |
NM_001038957.3 | NP_001034046.3 | 3 | 1543 | 78 | ||
NM_001038959.3 | NP_001034048.2 | 3 | 1543 | 78 | ||
NM_141953.3 | NP_650210.2 | 3 | 1543 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |