Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079124.3 | NP_523848.2 | 3 | 840 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |