Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Argk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104085.2 | NP_001097555.1 | 3 | 1687 | 96 | |
NM_001104086.2 | NP_001097556.2 | 3 | 1943 | 96 | ||
NM_001300060.1 | NP_001286989.1 | 1 | 1310 | 96 | ||
NM_079264.3 | NP_523988.2 | 2 | 1352 | 96 | ||
NM_168311.3 | NP_729446.1 | 4 | 2248 | 96 | ||
NM_168312.3 | NP_729447.1 | 3 | 2011 | 96 | ||
NM_168313.4 | NP_729448.1 | 2 | 2085 | 96 | ||
GenBank mRNA | AY069099.1 | - | 4 | 2050 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |