Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pHCl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038934.4 | NP_001034023.2 | 8 - 9 | 1877 | 62 | |
NM_001038935.3 | NP_001034024.2 | 9 - 10 | 1955 | 62 | ||
NM_001038936.4 | NP_001034025.2 | 9 - 10 | 1952 | 62 | ||
NM_001038937.2 | NP_001034026.1 | 9 - 10 | 1955 | 62 | ||
NM_001169979.1 | NP_001163450.1 | 9 - 10 | 1955 | 62 | ||
NM_001259852.1 | NP_001246781.1 | 9 - 10 | 1960 | 62 | ||
NM_001259853.1 | NP_001246782.1 | 8 - 9 | 1885 | 62 | ||
NM_001274965.1 | NP_001261894.1 | 8 - 9 | 1939 | 62 | ||
NM_001274966.1 | NP_001261895.1 | 7 - 8 | 1864 | 62 | ||
NM_001274967.1 | NP_001261896.1 | 7 - 8 | 1864 | 62 | ||
NM_001274968.1 | NP_001261897.1 | 8 - 9 | 1934 | 62 | ||
NM_001274969.1 | NP_001261898.1 | 9 - 10 | 1955 | 62 | ||
NM_001274970.1 | NP_001261899.1 | 7 - 8 | 1859 | 62 | ||
NM_001274971.1 | NP_001261900.1 | 8 - 9 | 1934 | 62 | ||
NM_001370024.1 | NP_001356981.1 | 8 - 9 | 1939 | 62 | ||
NM_001370025.1 | NP_001356910.1 | 9 - 10 | 1960 | 62 | ||
NM_001370026.1 | NP_001356909.1 | 7 - 8 | 1864 | 62 | ||
GenBank mRNA | BT012472.1 | - | 9 - 10 | 1956 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |