Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skeletor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032001.2 | NP_001027172.1 | 3 | 1036 | 148 | |
GenBank mRNA | AF321290.1 | - | 3 | 1017 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |