Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppr-Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015502.3 | NP_001015502.3 | 1 - 2 | 195 | 108 | |
GenBank mRNA | AF427495.1 | - | 1 - 2 | 195 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |