Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042968.2 | NP_001036433.1 | 1 - 2 | 183 | 108 | |
NM_001273770.2 | NP_001260699.1 | 1 - 2 | 183 | 108 | ||
GenBank mRNA | AY118852.1 | - | 1 - 2 | 131 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |