Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043176.2 | NP_001036641.1 | 5 - 6 | 433 | 112 | |
NM_001043177.3 | NP_001036642.2 | 5 - 6 | 433 | 112 | ||
GenBank mRNA | BT023789.1 | - | 5 - 6 | 384 | 112 | |
BT126217.1 | - | 5 - 6 | 385 | 112 | ||
L22021.1 | - | 5 - 6 | 382 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |