Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001814.1 | NP_001001814.1 | 1 - 2 | 140 | 61 | |
XM_009296614.2 | XP_009294889.1 | 2 - 3 | 204 | 61 | ||
GenBank mRNA | AY576995.1 | - | 1 - 2 | 140 | 61 | |
BC079518.1 | - | 1 - 2 | 109 | 61 | ||
BC148167.1 | - | 2 - 3 | 147 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |