Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 75 - 76 | 16190 | 63 | |
XM_005165511.4 | XP_005165568.1 | 75 - 76 | 16309 | 63 | ||
XM_009301753.3 | XP_009300028.1 | 76 - 77 | 16331 | 63 | ||
GenBank mRNA | AY540131.1 | - | 75 - 76 | 16194 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |