Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 43 - 44 | 9711 | 62 | |
XM_005165511.4 | XP_005165568.1 | 43 - 44 | 9830 | 62 | ||
XM_009301753.3 | XP_009300028.1 | 44 - 45 | 9852 | 62 | ||
GenBank mRNA | AY540131.1 | - | 43 - 44 | 9715 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |