Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cep43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002125.1 | NP_001002125.1 | 14 - 15 | 1183 | 95 | |
XM_005160662.4 | XP_005160719.1 | 15 - 16 | 1280 | 95 | ||
XM_021468390.1 | XP_021324065.1 | 16 - 17 | 1355 | 95 | ||
XM_021468391.1 | XP_021324066.1 | 16 - 17 | 1355 | 95 | ||
XM_021468392.1 | XP_021324067.1 | 15 - 16 | 1292 | 95 | ||
XM_021468393.1 | XP_021324068.1 | 14 - 15 | 1211 | 95 | ||
GenBank mRNA | BC071450.1 | - | 14 - 15 | 1183 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |