Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpdu1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004545.1 | NP_001004545.1 | 4 - 5 | 488 | 78 | |
GenBank mRNA | BC081598.1 | - | 4 - 5 | 488 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |