Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2caa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017886.1 | NP_001017886.1 | 5 - 6 | 1082 | 72 | |
GenBank mRNA | BC092961.1 | - | 5 - 6 | 1082 | 72 | |
BC155238.1 | - | 5 - 6 | 1012 | 72 | ||
BC165383.1 | - | 5 - 6 | 765 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |