Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: noc4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024397.1 | NP_001019568.1 | 1 - 2 | 161 | 82 | |
GenBank mRNA | BC095257.1 | - | 1 - 2 | 161 | 82 | |
BC165709.1 | - | 1 - 2 | 176 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |