Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: igsf21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034184.2 | NP_001029356.2 | 5 - 6 | 797 | 86 | |
NM_001278321.1 | NP_001265250.1 | 5 - 6 | 797 | 86 | ||
GenBank mRNA | BC100052.1 | - | 5 - 6 | 797 | 86 | |
CU638803.1 | - | 5 - 6 | 616 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |