Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tamm41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037119.1 | NP_001032196.1 | 502 | 106 | ||
GenBank mRNA | BC107608.1 | - | 502 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio |