Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkacaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082840.1 | NP_001076309.1 | 4 - 5 | 906 | 87 | |
XM_021470955.1 | XP_021326630.1 | 4 - 5 | 665 | 87 | ||
GenBank mRNA | BC133159.1 | - | 4 - 5 | 906 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |