Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alcama | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131000.1 | NP_571075.1 | 9 - 10 | 1237 | 121 | |
XM_005172635.4 | XP_005172692.1 | 9 - 10 | 1263 | 121 | ||
GenBank mRNA | BC050482.1 | - | 9 - 10 | 1173 | 121 | |
BC071276.1 | - | 9 - 10 | 1238 | 121 | ||
CU458879.1 | - | 9 - 10 | 1078 | 121 | ||
CU458962.1 | - | 9 - 10 | 1184 | 121 | ||
L25273.1 | - | 9 - 10 | 1237 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |