Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spon1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131515.1 | NP_571590.1 | 8 - 9 | 1246 | 107 | |
GenBank mRNA | AB006086.1 | - | 8 - 9 | 1246 | 107 | |
BC162586.1 | - | 8 - 9 | 1229 | 107 | ||
KM655720.1 | - | 8 - 9 | 1094 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |