Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: baiap2l1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198922.1 | NP_944604.1 | 7 - 8 | 793 | 81 | |
XM_009306593.3 | XP_009304868.1 | 7 - 8 | 832 | 81 | ||
GenBank mRNA | AY398378.1 | - | 7 - 8 | 794 | 81 | |
BC050238.1 | - | 7 - 8 | 793 | 81 | ||
BC068330.1 | - | 7 - 8 | 777 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |