Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam49ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291713.1 | NP_001278642.1 | 6 - 7 | 910 | 83 | |
NM_200070.3 | NP_956364.2 | 6 - 7 | 910 | 83 | ||
GenBank mRNA | AY394961.1 | - | 6 - 7 | 610 | 83 | |
BC053261.1 | - | 6 - 7 | 596 | 83 | ||
EH507686.1 | - | 2 - 3 | 311 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |