Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam49ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200070.3 | NP_956364.2 | 7 - 8 | 1029 | 80 | |
GenBank mRNA | AY394961.1 | - | 7 - 8 | 729 | 80 | |
EH507686.1 | - | 3 - 4 | 430 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |