Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200334.1 | NP_956628.1 | 7 - 8 | 880 | 55 | |
GenBank mRNA | BC053111.1 | - | 7 - 8 | 880 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |