Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2nb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200342.1 | NP_956636.1 | 2 - 3 | 355 | 68 | |
GenBank mRNA | BC053141.1 | - | 2 - 3 | 355 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |