Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200675.2 | NP_956969.1 | 6 - 7 | 541 | 69 | |
XM_005167296.4 | XP_005167353.2 | 6 - 7 | 623 | 69 | ||
XM_005167298.4 | XP_005167355.2 | 6 - 7 | 608 | 69 | ||
XM_005167299.4 | XP_005167356.1 | 6 - 7 | 628 | 69 | ||
GenBank mRNA | AY398406.1 | - | 6 - 7 | 541 | 69 | |
BC050162.1 | - | 6 - 7 | 633 | 69 | ||
BC058292.1 | - | 6 - 7 | 543 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |