Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem38a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200900.1 | NP_957194.1 | 5 - 6 | 716 | 70 | |
GenBank mRNA | BC064309.1 | - | 5 - 6 | 716 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |