Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200943.3 | NP_957237.1 | 5 - 6 | 720 | 124 | |
GenBank mRNA | BC055240.1 | - | 5 - 6 | 691 | 124 | |
EH447839.1 | - | 5 - 6 | 722 | 124 | ||
EH457748.1 | - | 5 - 6 | 724 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |