Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205703.1 | NP_991266.1 | 1577 | 103 | ||
GenBank mRNA | BC065962.1 | - | 1577 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio |