Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212609.1 | NP_997774.1 | 10 - 11 | 1380 | 119 | |
GenBank mRNA | BC056708.1 | - | 10 - 11 | 1380 | 119 | |
BC071277.1 | - | 10 - 11 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |