Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351837.1 | NP_001338766.1 | 12 - 13 | 2056 | 80 | |
NM_212779.2 | NP_997944.2 | 12 - 13 | 2056 | 80 | ||
XM_021479113.1 | XP_021334788.1 | 12 - 13 | 2119 | 80 | ||
GenBank mRNA | AF326958.1 | - | 1 - 2 | 193 | 80 | |
AF349034.1 | - | 12 - 13 | 2057 | 80 | ||
AF378824.1 | - | 9 - 10 | 1733 | 80 | ||
AF378825.1 | - | 3 - 4 | 523 | 80 | ||
AF378826.1 | - | 3 - 4 | 542 | 80 | ||
BC056287.1 | - | 12 - 13 | 2032 | 80 | ||
BC068379.1 | - | 12 - 13 | 2031 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |