Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gyg1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002062.1 | NP_001002062.1 | 2 - 3 | 189 | 63 | |
XM_009297232.3 | XP_009295507.1 | 2 - 3 | 214 | 63 | ||
GenBank mRNA | BC071363.1 | - | 2 - 3 | 189 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |