Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dus4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003490.2 | NP_001003490.2 | 2 - 3 | 416 | 77 | |
GenBank mRNA | BC078413.1 | - | 2 - 3 | 389 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |