Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003980.2 | NP_001003980.1 | 3 - 4 | 445 | 90 | |
XM_005171154.4 | XP_005171211.1 | 3 - 4 | 90 | |||
GenBank mRNA | AY664404.1 | - | 3 - 4 | 446 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |