Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sccpdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005574.1 | NP_001005574.1 | 2 - 3 | 515 | 89 | |
GenBank mRNA | BC066701.1 | - | 2 - 3 | 300 | 89 | |
BC083193.1 | - | 2 - 3 | 515 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |