Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sccpdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005574.1 | NP_001005574.1 | 9 - 10 | 1204 | 136 | |
GenBank mRNA | BC066701.1 | - | 9 - 10 | 989 | 136 | |
BC083193.1 | - | 9 - 10 | 1204 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |