Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: extl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008402.1 | NP_001008402.1 | 4 - 5 | 2633 | 64 | |
XM_005170211.4 | XP_005170268.1 | 5 - 6 | 3143 | 64 | ||
GenBank mRNA | AY786436.1 | - | 4 - 5 | 2633 | 64 | |
BC162406.1 | - | 4 - 5 | 2624 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |