Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nadka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017580.1 | NP_001017580.1 | 675 | 99 | ||
GenBank mRNA | BC092723.1 | - | 675 | 99 | ||
BC165884.1 | - | 525 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio |