Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyb5d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017684.2 | NP_001017684.1 | 2 - 3 | 281 | 65 | |
GenBank mRNA | BC093157.1 | - | 2 - 3 | 284 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |