Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017877.1 | NP_001017877.1 | 1 - 2 | 250 | 85 | |
GenBank mRNA | BC092971.1 | - | 1 - 2 | 250 | 85 | |
BC165377.1 | - | 1 - 2 | 112 | 85 | ||
KT008426.1 | - | 1 - 2 | 78 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |