Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hivep2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030164.1 | NP_001025335.1 | 2 - 3 | 5017 | 69 | |
XM_005160730.4 | XP_005160787.1 | 5 - 6 | 5954 | 69 | ||
XM_005160731.4 | XP_005160788.1 | 6 - 7 | 6026 | 69 | ||
XM_005160733.4 | XP_005160790.1 | 5 - 6 | 5823 | 69 | ||
XM_005160735.4 | XP_005160792.1 | 4 - 5 | 6688 | 69 | ||
XM_021468461.1 | XP_021324136.1 | 6 - 7 | 5895 | 69 | ||
XM_021468462.1 | XP_021324137.1 | 4 - 5 | 5534 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |