Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044396.2 | NP_001037861.1 | 1 - 2 | 266 | 77 | |
GenBank mRNA | AY865571.1 | - | 1 - 2 | 124 | 77 | |
BC076165.1 | - | 1 - 2 | 262 | 77 | ||
BC122138.1 | - | 1 - 2 | 266 | 77 | ||
BC165155.1 | - | 1 - 2 | 158 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |