Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myt1la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044899.2 | NP_001038364.1 | 3 - 4 | 142 | 96 | |
XM_005160722.4 | XP_005160779.1 | 5 - 6 | 513 | 96 | ||
XM_005160723.4 | XP_005160780.1 | 5 - 6 | 513 | 96 | ||
XM_009294757.3 | XP_009293032.1 | 5 - 6 | 513 | 96 | ||
GenBank mRNA | BC124350.1 | - | 5 - 6 | 472 | 96 | |
BC171626.1 | - | 4 - 5 | 211 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |