Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdyhv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044947.1 | NP_001038412.1 | 2 - 3 | 184 | 136 | |
GenBank mRNA | BC122321.1 | - | 1 - 2 | 119 | 136 | |
BC171472.1 | - | 2 - 3 | 238 | 136 | ||
BC171474.1 | - | 2 - 3 | 238 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |