Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loxl5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045856.2 | NP_001039321.2 | 2 - 3 | 951 | 57 | |
XM_021469354.1 | XP_021325029.1 | 2 - 3 | 1055 | 57 | ||
GenBank mRNA | EF030485.1 | - | 2 - 3 | 951 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |