Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loxl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099244.1 | NP_001092714.1 | 3 - 4 | 650 | 104 | |
XM_009305250.3 | XP_009303525.1 | 4 - 5 | 836 | 104 | ||
GenBank mRNA | EF030481.1 | - | 3 - 4 | 650 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |